Please login to your online account to display your discounted pricing

Ethyl N-ethylcarbamate, ACROS Organics™


Quantity 10g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC119070100

 View more versions of this product

Catalog No. AC119070100

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 117.15
Quantity 10g
Color Clear slightly yellow
Beilstein 04, 114
Density 0.9810g/mL
Linear Formula C2H5NHCO2C2H5
CAS 623-78-9
MDL Number MFCD00009102
Packaging Glass bottle
Infrared Spectrum Authentic
Synonym N-Ethylurethane
Refractive Index 1.4250 to 1.4270 (20°C, 589nm)
Assay Percent Range 98%
Chemical Name or Material Ethyl N-ethylcarbamate
Molecular Formula C5H11NO2
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Health Hazard 3 GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Ethyl N-ethylcarbamate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H11NO2
Linear Formula C2H5NHCO2C2H5
Formula Weight 117.15
MDL Number MFCD00009102
Synonym N-Ethylurethane
Accessories

GHS P Statement:
Use personal protective equipment as required.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.

SDS & Safety Documents