missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 97%, tech., Acros Organics

Chemical Identifiers

CAS 52763-21-0
Molecular Formula C15H19NO3·HCl
Molecular Weight (g/mol) 297.77
MDL Number MFCD00012792
InChI Key UQOMEAWPKSISII-UHFFFAOYSA-N
Synonym 1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride
PubChem CID 2723880
IUPAC Name ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride
SMILES CCOC(=O)C1CCN(CC1=O)CC2=CC=CC=C2.Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC162300050 SDS View Documents Thermo Fisher Scientific
162300050
5g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC162300250 SDS View Documents Thermo Fisher Scientific
162300250
25g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC162301000 SDS View Documents Thermo Fisher Scientific
AC162301000
100g Glass bottle N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

52763-21-0
297.77
UQOMEAWPKSISII-UHFFFAOYSA-N
2723880
CCOC(=O)C1CCN(CC1=O)CC2=CC=CC=C2.Cl
C15H19NO3·HCl
MFCD00012792
1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride

Specifications

Authentic
White to Brown
5g
97%
MFCD00012792
UQOMEAWPKSISII-UHFFFAOYSA-N
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride
2723880
Crystalline Powder
Technical
Glass bottle
160°C to 165°C
52763-21-0
C15H19NO3·HCl
1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride
CCOC(=O)C1CCN(CC1=O)CC2=CC=CC=C2.Cl
297.77
297.77
97%
Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 258-162-5