missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 97%, tech., ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 162300250

 View more versions of this product

Catalog No. AC162300250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride
Glass bottle
160°C to 165°C
52763-21-0
C15H19NO3·HCl
1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, pubchem19671, zlchem 420
CCOC(=O)C1CCN(CC1=O)CC2=CC=CC=C2.Cl
297.77
297.77
97%
Authentic
White to Brown
25g
97%
MFCD00012792
UQOMEAWPKSISII-UHFFFAOYSA-N
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride
2723880
Crystalline Powder
Technical

Chemical Identifiers

52763-21-0
297.77
UQOMEAWPKSISII-UHFFFAOYSA-N
2723880
CCOC(=O)C1CCN(CC1=O)CC2=CC=CC=C2.Cl
C15H19NO3·HCl
MFCD00012792
1-benzyl-3-oxopiperidine-4-carboxylic acid ethyl ester hydrochloride, 1-benzyl-4-ethoxycarbonyl-3-piperidone hydrochloride, ethyl 1-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hcl, ethyl 1-benzyl-3-oxo-piperidine-4-carboxylate hydrochloride, ethyl 1-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, ethyl n-benzyl-3-oxo-4-piperidine-carboxylate hydrochloride, ethyl n-benzyl-3-oxopiperidine-4-carboxylate hydrochloride, pubchem19671, zlchem 420
ethyl 1-benzyl-3-oxopiperidine-4-carboxylate;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 258-162-5

SDS Documents