Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl N-(4-bromophenyl)oxamate

$163.16 - $726.54

Chemical Identifiers

Chemical Name or Material Ethyl N-(4-bromophenyl)oxamate
CAS 24451-15-8
Molecular Formula C10H10BrNO3
Formula Weight 272.1
MDL Number MFCD00728542
Synonym N-(4-Bromophenyl)oxamic acid ethyl ester; Ethyl (4-bromoanilino)oxoacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5059803

 
alfa aesar™
H5059803
1g Each for $163.16

AAH5059806

 
alfa aesar™
H5059806
5g Each for $726.54
Description & Specifications

Specifications

Formula Weight 272.1
Chemical Name or Material Ethyl N-(4-bromophenyl)oxamate
Molecular Formula C10H10BrNO3
MDL Number MFCD00728542
CAS 24451-15-8
Synonym N-(4-Bromophenyl)oxamic acid ethyl ester; Ethyl (4-bromoanilino)oxoacetate
Quantity 1g