Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl N-(2,4-dinitrophenoxy)acetimidate, 98+%

$77.15 - $679.55

Chemical Identifiers

Chemical Name or Material Ethyl N-(2,4-dinitrophenoxy)acetimidate
CAS 54322-32-6
Molecular Formula C10H11N3O6
Formula Weight 269.21
MDL Number MFCD00010133
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2634103

  SDS
alfa aesar™
H2634103
1g Each for $77.15

AAH2634106

  SDS
alfa aesar™
H2634106
5g Each for $244.09

AAH2634114

  SDS
alfa aesar™
H2634114
25g Each for $679.55
Description & Specifications

Specifications

Formula Weight 269.21
MDL Number MFCD00010133
Quantity 1g
Health Hazard 1 Warning
CAS 54322-32-6
Molecular Formula C10H11N3O6
Assay ≥98%
Melting Point 110° to 113°C
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Ethyl N-(2,4-dinitrophenoxy)acetimidate
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.