missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl myristate, 98+%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 385541000

 View more versions of this product

Catalog No. AC385541000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethyl myristate
0.8610g/mL
>110°C
98% min. (GC)
C16H32O2
MFCD00008984
0.861
ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
MMKRHZKQPFCLLS-UHFFFAOYSA-N
ethyl tetradecanoate
31283
256.43
124-06-1
295.0°C
Authentic
Glass bottle
CH3(CH2)12COOCH2CH3
100g
15, 6419
Solubility in water: insoluble
CCCCCCCCCCCCCC(=O)OCC
256.43
CHEBI:84849
98+%

Chemical Identifiers

124-06-1
256.43
MMKRHZKQPFCLLS-UHFFFAOYSA-N
31283
ethyl tetradecanoate
C16H32O2
MFCD00008984
ethyl myristate, tetradecanoic acid, ethyl ester, myristic acid ethyl ester, myristic acid, ethyl ester, ethylmyristate, ethyl n-tetradecanoate, ethyl myristate natural, fema no. 2445, tetradecanoic acid ethyl ester, ethyl ester tetradecanoic acid
CHEBI:84849
CCCCCCCCCCCCCC(=O)OCC
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 204-675-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only