Learn More
Ethyl morpholinoacetate, 98%, Thermo Scientific™
$46.78 - $46.78
Chemical Identifiers
CAS | 3235-82-3 |
---|---|
Molecular Formula | C8H15NO3 |
Molecular Weight (g/mol) | 173.21 |
MDL Number | MFCD00023375 |
InChI Key | SITMDWHJQROIPF-UHFFFAOYSA-N |
Synonym | ethyl 2-morpholinoacetate, 4-morpholineacetic acid, ethyl ester, ethyl morpholinoacetate, ethyl 2-morpholin-4-yl acetate, morpholin-4-yl-acetic acid ethyl ester, ethyl 4-morpholineacetate, ethyl-2-morpholinoacetate, ethyl 4-morpholinylacetate, ethyl morpholin-4-ylacetate, morpholinoacetic acid ethyl ester |
PubChem CID | 2734105 |
IUPAC Name | ethyl 2-morpholin-4-ylacetate |
SMILES | CCOC(=O)CN1CCOCC1 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC350910050
|
Thermo Scientific Chemicals
350910050 |
5 g | Glass bottle |
Each for $46.78
|
|
||||
Chemical Identifiers
3235-82-3 | |
173.21 | |
SITMDWHJQROIPF-UHFFFAOYSA-N | |
2734105 | |
CCOC(=O)CN1CCOCC1 |
C8H15NO3 | |
MFCD00023375 | |
ethyl 2-morpholinoacetate, 4-morpholineacetic acid, ethyl ester, ethyl morpholinoacetate, ethyl 2-morpholin-4-yl acetate, morpholin-4-yl-acetic acid ethyl ester, ethyl 4-morpholineacetate, ethyl-2-morpholinoacetate, ethyl 4-morpholinylacetate, ethyl morpholin-4-ylacetate, morpholinoacetic acid ethyl ester | |
ethyl 2-morpholin-4-ylacetate |
Specifications
3235-82-3 | |
Authentic | |
Glass bottle | |
1.4535 to 1.4555 | |
5 g | |
SITMDWHJQROIPF-UHFFFAOYSA-N | |
ethyl 2-morpholin-4-ylacetate | |
2734105 | |
98% | |
Ethyl morpholinoacetate, 98% |
Yellow | |
97.5% min. (GC) | |
C8H15NO3 | |
MFCD00023375 | |
ethyl 2-morpholinoacetate, 4-morpholineacetic acid, ethyl ester, ethyl morpholinoacetate, ethyl 2-morpholin-4-yl acetate, morpholin-4-yl-acetic acid ethyl ester, ethyl 4-morpholineacetate, ethyl-2-morpholinoacetate, ethyl 4-morpholinylacetate, ethyl morpholin-4-ylacetate, morpholinoacetic acid ethyl ester | |
CCOC(=O)CN1CCOCC1 | |
173.21 | |
173.21 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
RUO – Research Use Only