Learn More
Ethyl (methylthio)acetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2428514
Description
Ethyl (methylthio)acetate is used as an internal standard during the analytical determination of volatile sulfur compounds in port wines. Further, it is used as food flavors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl (methylthio)acetate is used as an internal standard during the analytical determination of volatile sulfur compounds in port wines. Further, it is used as food flavors.
Solubility
Miscible with alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong alkalis.
Specifications
Ethyl (methylthio)acetate | |
1.06 | |
59°C (138°F) | |
C5H10O2S | |
MFCD00009182 | |
UN3272 | |
ethyl methylthio acetate, ethyl 2-methylthio acetate, acetic acid, methylthio-, ethyl ester, ethyl 2-methylthio-acetate, ethyl 2-methylsulfanyl acetate, unii-6148qqn25t, ethyl alpha-methylthio acetate, methylthio acetic acid ethyl ester, ethyl .alpha. methylthio acetate, methylsulfanyl-acetic acid ethyl ester | |
MDIAKIHKBBNYHF-UHFFFAOYSA-N | |
ethyl 2-methylsulfanylacetate | |
78199 | |
98% |
4455-13-4 | |
176°C | |
Characteristic | |
1.46 | |
25 g | |
1744999 | |
Miscible with alcohol. Immiscible with water. | |
CCOC(=O)CSC | |
134.193 | |
134.2 |
Chemical Identifiers
4455-13-4 | |
134.193 | |
MDIAKIHKBBNYHF-UHFFFAOYSA-N | |
78199 | |
CCOC(=O)CSC |
C5H10O2S | |
MFCD00009182 | |
ethyl methylthio acetate, ethyl 2-methylthio acetate, acetic acid, methylthio-, ethyl ester, ethyl 2-methylthio-acetate, ethyl 2-methylsulfanyl acetate, unii-6148qqn25t, ethyl alpha-methylthio acetate, methylthio acetic acid ethyl ester, ethyl .alpha. methylthio acetate, methylsulfanyl-acetic acid ethyl ester | |
ethyl 2-methylsulfanylacetate |
Safety and Handling
GHS H Statement
H226-H319-H335
Flammable liquid and vapour.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 224-700-2
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only