Please login to your online account to display your discounted pricing

Ethyl (methylthio)acetate, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  198120250

 View more versions of this product

Catalog No. AC198120250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 134.19
Physical Form Liquid
Quantity 25g
Color Clear colorless to slightly yellow
EINECS Number 224-700-2
Density 1.04g/mL
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Infrared Spectrum Authentic
Refractive Index 1.4577 to 1.4597 (20°C, 589nm)
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CH3SCH2CO2C2H5
MDL Number MFCD00009182
Chemical Name or Material Ethyl (methylthio)acetate
Molecular Formula C5H10O2S
Assay Percent Range 95%
Name Note 95%
CAS 4455-13-4

Specifications

Chemical Name or Material Ethyl (methylthio)acetate
Name Note 95%
CAS 4455-13-4
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H10O2S
Linear Formula CH3SCH2CO2C2H5
Formula Weight 134.19
MDL Number MFCD00009182
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.

SDS & Safety Documents