Please login to your online account to display your discounted pricing

Ethyl (methylthio)acetate, 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl (methylthio)acetate
Name Note 95%
CAS 4455-13-4
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H10O2S
Linear Formula CH3SCH2CO2C2H5
Formula Weight 134.19
MDL Number MFCD00009182
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC198120250

  SDS
thermo fisher scientific
198120250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC198121000

  SDS
thermo fisher scientific
198121000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 134.19
Quantity 25g
Color Clear colorless to slightly yellow
EINECS Number 224-700-2
Density 1.04g/mL
Chemical Name or Material Ethyl (methylthio)acetate
Packaging Glass bottle
Refractive Index 1.4577 to 1.4597 (20°C, 589nm)
Name Note 95%
Assay Percent Range 95%
Molecular Formula C5H10O2S
Linear Formula CH3SCH2CO2C2H5
MDL Number MFCD00009182
CAS 4455-13-4
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.