Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl methylsulfonylacetate, 97%

$107.59 - $107.59

Chemical Identifiers

Chemical Name or Material Ethyl methylsulfonylacetate
CAS 4455-15-6
Molecular Formula C5H10O4S
Formula Weight 166.2
MDL Number MFCD00041258
Beilstein 1771631
Synonym Ethyl methanesulfonylacetate; Methylsulfonylacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0507706

  SDS
alfa aesar™
L0507706
5g Each for $107.59
Description & Specifications

Specifications

Formula Weight 166.2
Chemical Name or Material Ethyl methylsulfonylacetate
CAS 4455-15-6
MDL Number MFCD00041258
Assay 97%
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Health Hazard 1 Warning
Beilstein 1771631
Synonym Ethyl methanesulfonylacetate; Methylsulfonylacetic acid ethyl ester
Flash Point >100°C (212°F)
Quantity 5g
Molecular Formula C5H10O4S
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Boiling Point 111° to 113°C (0.2mmHg)
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.