Please login to your online account to display your discounted pricing

Ethyl methoxyacetate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl methoxyacetate
Name Note 98%
CAS 3938-96-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H10O3
Linear Formula CH3OCH2CO2C2H5
Formula Weight 118.13
MDL Number MFCD00009176
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC214040500

  SDS
thermo fisher scientific
214040500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 118.13
Quantity 50g
Color Clear colorless
EINECS Number 223-521-7
Density 1.0000g/mL
Beilstein 03, 236
Chemical Name or Material Ethyl methoxyacetate
Packaging Glass bottle
Refractive Index 1.3985 to 1.4015 (20°C, 589nm)
Additional Information product may contain variable amounts of ethanol
Name Note 98%
Assay Percent Range 98%
Molecular Formula C5H10O3
Linear Formula CH3OCH2CO2C2H5
MDL Number MFCD00009176
CAS 3938-96-3
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.
Highly flammable liquid and vapor.