Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-Ethyl mandelate, 97%

$59.38 - $162.73

Chemical Identifiers

Chemical Name or Material (+/-)-Ethyl mandelate
CAS 774-40-3
Molecular Formula C10H12O3
Formula Weight 180.2
MDL Number MFCD00004494
Synonym Ethyl alpha-hydroxyphenylacetate; Mandelic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1741214

  SDS
alfa aesar™
A1741214
25g Each for $59.38

AAA1741222

  SDS
alfa aesar™
A1741222
100g Each for $162.73
Description & Specifications

Specifications

Formula Weight 180.2
Chemical Name or Material (+/-)-Ethyl mandelate
Assay 97%
Molecular Formula C10H12O3
MDL Number MFCD00004494
CAS 774-40-3
Synonym Ethyl alpha-hydroxyphenylacetate; Mandelic acid ethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H319
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
RTECS Number OO7397000
Flash Point >110°C (230°F)
Boiling Point 253° to 255°C
Quantity 25g
Safety and Handling

Warning

GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.

GHS H Statement
H319
Causes serious eye irritation.