missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Malonyl Chloride, TCI America™

Manufacturer:  TCI America E048425G

 View more versions of this product

Catalog No. E048425G



Specifications

Specifications

Ethyl Malonyl Chloride
Yellow
36239-09-5
MFCD00000736
ethyl 3-chloro-3-oxopropionate, ethyl chloroformylacetate, ethyl chloroformyl acetate, propanoic acid, 3-chloro-3-oxo-, ethyl ester, ethyl3-chloro-3-oxopropanoate, ethyl malonylchloride, carbethoxyacetyl chloride, 2-ethoxycarbonylacetyl chloride, ethyl 2-chlorocarbonyl acetate, chloroformyl acetic acid ethyl ester
CCOC(=O)CC(=O)Cl
150.558
150.56
≥97.0% (GC,T)
83°C
25g
C5H7ClO3
3265
KWFADUNOPOSMIJ-UHFFFAOYSA-N
ethyl 3-chloro-3-oxopropanoate
118931
Clear Liquid at 20°C

Chemical Identifiers

36239-09-5
150.558
KWFADUNOPOSMIJ-UHFFFAOYSA-N
118931
CCOC(=O)CC(=O)Cl
C5H7ClO3
MFCD00000736
ethyl 3-chloro-3-oxopropionate, ethyl chloroformylacetate, ethyl chloroformyl acetate, propanoic acid, 3-chloro-3-oxo-, ethyl ester, ethyl3-chloro-3-oxopropanoate, ethyl malonylchloride, carbethoxyacetyl chloride, 2-ethoxycarbonylacetyl chloride, ethyl 2-chlorocarbonyl acetate, chloroformyl acetic acid ethyl ester
ethyl 3-chloro-3-oxopropanoate
Safety and Handling

Safety and Handling

TSCA : No

Documents