Please login to your online account to display your discounted pricing

Ethyl malonyl chloride, 90%, tech., ACROS Organics™

$69.28 - $221.31

Chemical Identifiers

Chemical Name or Material Ethyl malonyl chloride
Assay ≥88% (RCOCL)
Assay Percent Range 90%
Molecular Formula C5H7ClO3
Linear Formula ClCOCH2CO2C2H5
Formula Weight 150.56
Synonym Ethyl (chloroformyl)acetate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC172690050

  SDS
thermo fisher scientific
172690050
Glass bottle 5g Each for $69.28

AC172690250

  SDS
thermo fisher scientific
172690250
Glass bottle 25g Each for $221.31

AC172691000

  SDS
thermo fisher scientific
AC172691000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Ethyl malonyl chloride
Color Clear yellow to brown
Quantity 5g
CAS 36239-09-5
Density 1.1700g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4250 to 1.4310 (20°C, 589nm)
Physical Form Liquid
Assay ≥88% (RCOCL)
Assay Percent Range 90%
Molecular Formula C5H7ClO3
Linear Formula ClCOCH2CO2C2H5
Formula Weight 150.56
Synonym Ethyl (chloroformyl)acetate