Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl m-tolylacetate, 98%

$80.35 - $260.14

Chemical Identifiers

Chemical Name or Material Ethyl m-tolylacetate
CAS 40061-55-0
Molecular Formula C11H14O2
Formula Weight 178.23
MDL Number MFCD00017273
Beilstein 2252144
Synonym m-Tolylacetic acid ethyl ester; Ethyl 3-methylphenylacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1849709

 
alfa aesar™
A1849709
10g Each for $80.35

AAA1849718

 
alfa aesar™
A1849718
50g Each for $260.14
Description & Specifications

Specifications

Formula Weight 178.23
Chemical Name or Material Ethyl m-tolylacetate
Assay 98%
Molecular Formula C11H14O2
MDL Number MFCD00017273
CAS 40061-55-0
Beilstein 2252144
Synonym m-Tolylacetic acid ethyl ester; Ethyl 3-methylphenylacetate
Flash Point >110°C (230°F)
Boiling Point 237° to 238°C
Quantity 10g