Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl m-toluate, 98+%

$32.95 - $84.97

Chemical Identifiers

Chemical Name or Material Ethyl m-toluate
CAS 120-33-2
Molecular Formula C10H12O2
Formula Weight 164.21
MDL Number MFCD00009114
Beilstein 1863739
Synonym Ethyl 3-methylbenzoate; m-Toluic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2392514

 
alfa aesar™
B2392514
25g Each for $32.95

AAB2392522

 
alfa aesar™
B2392522
100g Each for $84.97
Description & Specifications

Specifications

Formula Weight 164.21
Quantity 25g
Chemical Name or Material Ethyl m-toluate
Assay ≥98%
Molecular Formula C10H12O2
MDL Number MFCD00009114
CAS 120-33-2
Beilstein 1863739
Synonym Ethyl 3-methylbenzoate; m-Toluic acid ethyl ester
Flash Point 101°C (213°F)
Boiling Point 103° to 105°C (10mmHg)