missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Linoleate, TCI America™

$44.31 - $333.06

Chemical Identifiers

CAS 544-35-4
Molecular Formula C20H36O2
Molecular Weight (g/mol) 308.506
MDL Number MFCD00009535
InChI Key FMMOOAYVCKXGMF-MURFETPASA-N
Synonym ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester
PubChem CID 5282184
ChEBI CHEBI:31572
IUPAC Name ethyl (9Z,12Z)-octadeca-9,12-dienoate
SMILES CCCCCC=CCC=CCCCCCCCC(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
L005525ML View Documents TCI America
L005525ML
25mL Each for $44.31
 
L0055500ML View Documents TCI America
L0055500ML
500mL Each for $333.06
 
Specifications

Chemical Identifiers

544-35-4
308.506
FMMOOAYVCKXGMF-MURFETPASA-N
5282184
ethyl (9Z,12Z)-octadeca-9,12-dienoate
C20H36O2
MFCD00009535
ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester
CHEBI:31572
CCCCCC=CCC=CCCCCCCCC(=O)OCC

Specifications

193°C
25mL
C20H36O2
ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester
CCCCCC=CCC=CCCCCCCCC(=O)OCC
308.506
CHEBI:31572
Clear Liquid at 20°C
Ethyl Linoleate
Yellow
544-35-4
MFCD00009535
FMMOOAYVCKXGMF-MURFETPASA-N
ethyl (9Z,12Z)-octadeca-9,12-dienoate
5282184
308.51
≥70.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

RTECSNumber : RG2185000

TSCA : Yes