missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Linoleate, TCI America™

Manufacturer:  TCI America L0055500ML

 View more versions of this product

Catalog No. L0055500ML



Specifications

Specifications

Ethyl Linoleate
Yellow
544-35-4
MFCD00009535
FMMOOAYVCKXGMF-MURFETPASA-N
ethyl (9Z,12Z)-octadeca-9,12-dienoate
5282184
308.51
≥70.0% (GC)
193°C
500mL
C20H36O2
ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester
CCCCCC=CCC=CCCCCCCCC(=O)OCC
308.506
CHEBI:31572
Clear Liquid at 20°C

Chemical Identifiers

544-35-4
308.506
FMMOOAYVCKXGMF-MURFETPASA-N
5282184
ethyl (9Z,12Z)-octadeca-9,12-dienoate
C20H36O2
MFCD00009535
ethyl linoleate, linoleic acid ethyl ester, mandenol, ethyl linolate, ethyl cis,cis-9,12-octadecadienoate, unii-mj2ytt4j8m, ethyl linoleate jan, mj2ytt4j8m, ethyl z,z-9,12-octadecadienoate, linoleic acid, ethyl ester
CHEBI:31572
CCCCCC=CCC=CCCCCCCCC(=O)OCC
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0798

RTECSNumber : RG2185000

TSCA : Yes

Documents