missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl levulinate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 153621000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl levulinate | |
539-88-8 | |
Yellow | |
206°C | |
Authentic | |
Glass bottle | |
1.4210 to 1.423 | |
MFCD00009209 | |
03, 675 | |
15, 3874 | |
Solubility in water: 40g/L(20°C). Other solubilities: miscible with alcohol | |
CCOC(=O)CCC(C)=O | |
144.17 | |
144.17 | |
Liquid |
98% | |
5mg KOH/g max. | |
1.0120g/mL | |
90°C | |
97.5% min. (GC) | |
C7H12O3 | |
CH3COCH2CH2CO2C2H5 | |
100 g | |
1.012 | |
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate | |
GMEONFUTDYJSNV-UHFFFAOYSA-N | |
ethyl 4-oxopentanoate | |
10883 | |
98% |
Chemical Identifiers
539-88-8 | |
144.17 | |
GMEONFUTDYJSNV-UHFFFAOYSA-N | |
10883 | |
CCOC(=O)CCC(C)=O |
C7H12O3 | |
MFCD00009209 | |
ethyl levulinate, ethyl levulate, ethyl laevulinate, levulinic acid, ethyl ester, ethyl 4-ketovalerate, ethyl 3-acetylpropionate, ethyl 4-oxovalerate, pentanoic acid, 4-oxo-, ethyl ester, ethyl ketovalerate, ethyl acetylpropanoate | |
ethyl 4-oxopentanoate |
Safety and Handling
EINECSNumber : 208-728-2
RTECSNumber : OI1700000
TSCA : TSCA
RUO – Research Use Only