missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl L-(+)-Mandelate, TCI America™

$108.47 - $369.73

Chemical Identifiers

CAS 13704-09-1
Molecular Formula C10H12O3
Molecular Weight (g/mol) 180.203
MDL Number MFCD00064249
InChI Key SAXHIDRUJXPDOD-VIFPVBQESA-N
Synonym L-(+)-Mandelic Acid Ethyl Ester
PubChem CID 6951557
ChEBI CHEBI:78405
IUPAC Name ethyl (2S)-2-hydroxy-2-phenylacetate
SMILES CCOC(=O)C(C1=CC=CC=C1)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M12735G View Documents TCI America
M12735G
5g Each for $108.47
 
M127325G View Documents TCI America
M127325G
25g Each for $369.73
 
Specifications

Chemical Identifiers

13704-09-1
180.203
SAXHIDRUJXPDOD-VIFPVBQESA-N
6951557
ethyl (2S)-2-hydroxy-2-phenylacetate
C10H12O3
MFCD00064249
L-(+)-Mandelic Acid Ethyl Ester
CHEBI:78405
CCOC(=O)C(C1=CC=CC=C1)O

Specifications

107°C
5g
C10H12O3
L-(+)-Mandelic Acid Ethyl Ester
CCOC(=O)C(C1=CC=CC=C1)O
180.203
CHEBI:78405
Crystal-Lump at 20°C
Ethyl L-(+)-Mandelate
White-Yellow
13704-09-1
MFCD00064249
SAXHIDRUJXPDOD-VIFPVBQESA-N
ethyl (2S)-2-hydroxy-2-phenylacetate
6951557
180.20
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3841

TSCA : No