missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl L(-)-lactate, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC118660025
Specifications
0.1% max. (as lactic acid) | |
1.0340g/mL | |
Authentic | |
1.4100 to 1.4160 | |
− 10.00 (20.00°C neat) | |
0.3% max. (K.F.) | |
Undesignated | |
2.5L | |
96% min. (GC) | |
CH3CH(OH)CO2C2H5 | |
03,264 | |
14,3817 | |
Solubility in water: soluble. Other solubilities: miscible with alcohols,ketones and esters | |
CCOC(=O)C(C)O | |
118.13 | |
CHEBI:78322 | |
Liquid |
Ethyl L(-)-lactate | |
46°C | |
Glass bottle | |
1.034 | |
− 10.00 | |
154.0°C | |
-26.0°C | |
687-47-8 | |
C5H10O3 | |
MFCD00004518 | |
17,135 | |
ethyl 2s-2-hydroxypropanoate, ethyl l---lactate, ethyl l--lactate, ethyl l-lactate, ethyl s-2-hydroxypropionate, l---ethyl lactate, l-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s, s-ethyl lactate | |
LZCLXQDLBQLTDK-BYPYZUCNSA-N | |
ethyl (2S)-2-hydroxypropanoate | |
92831 | |
118.13 | |
97% |
Chemical Identifiers
687-47-8 | |
118.13 | |
LZCLXQDLBQLTDK-BYPYZUCNSA-N | |
92831 | |
ethyl (2S)-2-hydroxypropanoate |
C5H10O3 | |
MFCD00004518 | |
ethyl 2s-2-hydroxypropanoate, ethyl l---lactate, ethyl l--lactate, ethyl l-lactate, ethyl s-2-hydroxypropionate, l---ethyl lactate, l-ethyl lactate, propanoic acid, 2-hydroxy-, ethyl ester, 2s, s-ethyl lactate | |
CHEBI:78322 | |
CCOC(=O)C(C)O |
Safety and Handling
GHS H Statement
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
GHS P Statement
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physici
GHS Signal Word: Danger
EINECSNumber : 211-694-1