Please login to your online account to display your discounted pricing

Ethyl L(-)-lactate 95%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  410165000

 View more versions of this product

Catalog No. AC410165000

  • / Each
 

Description & Specifications

Chemical Identifiers

Density 1.0340g/mL
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4110 to 1.4150 (20°C, 589nm)
Specific Rotation -8° to -12° (20°C, 589nm) (neat)
Specific Rotation Condition − 10.00 (20.00°C neat)
Color Clear colorless to pale yellow
Quantity 500g
Chemical Name or Material Ethyl L(-)-lactate
Name Note 95%
CAS 687-47-8
Physical Form Clear colorless to pale yellow liquid
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C5H10O3
Linear Formula CH3CH(OH)CO2C2H5
Formula Weight 118.13g/mol
MDL Number MFCD00004518
Beilstein 03, 264
Fieser 17,135
Merck Index 14, 3817
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 211-694-1
TSCA TSCA

Specifications

Chemical Name or Material Ethyl L(-)-lactate
Name Note 95%
CAS 687-47-8
Assay 94% min.
Assay Percent Range 95%
Molecular Formula C5H10O3
Linear Formula CH3CH(OH)CO2C2H5
Formula Weight 118.13g/mol
MDL Number MFCD00004518
Accessories

Danger

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.

SDS & Safety Documents