Please login to your online account to display your discounted pricing

Ethyl L(-)-lactate 95%, ACROS Organics™

$20.02 - $20.02

Chemical Identifiers

Chemical Name or Material Ethyl L(-)-lactate
Name Note 95%
CAS 687-47-8
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C5H10O3
Linear Formula CH3CH(OH)CO2C2H5
Formula Weight 118.13
MDL Number MFCD00004518
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC410161000

  SDS
thermo fisher scientific
410161000
Glass bottle 100g Each for $20.02
Description & Specifications

Specifications

Assay ≥94%
Physical Form Liquid
Formula Weight 118.13
Quantity 100g
Color Clear colorless to pale yellow
Beilstein 03, 264
Fieser 17,135
Merck Index 14, 3817
EINECS Number 211-694-1
TSCA TSCA
Density 1.0340g/mL
Specific Rotation Condition − 10.00 (20.00°C neat)
Packaging Glass bottle
Refractive Index 1.4110 to 1.4150 (20°C, 589nm)
Specific Rotation -8° to -12° (20°C, 589nm) (neat)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Chemical Name or Material Ethyl L(-)-lactate
Name Note 95%
Assay Percent Range 95%
Molecular Formula C5H10O3
Linear Formula CH3CH(OH)CO2C2H5
MDL Number MFCD00004518
CAS 687-47-8