missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl isovalerate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A10292AE

 View more versions of this product

Catalog No. AAA10292AE


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl isovalerate
-99°C
131°C to 133°C
Fruit-like
1.396
100 mL
1744677
ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
CCOC(=O)CC(C)C
130.187
CHEBI:31571
98%
108-64-5
0.866
26°C (78°F)
C7H14O2
MFCD00009203
UN3272
14,3816
PPXUHEORWJQRHJ-UHFFFAOYSA-N
ethyl 3-methylbutanoate
7945
130.19

Chemical Identifiers

108-64-5
130.187
PPXUHEORWJQRHJ-UHFFFAOYSA-N
7945
ethyl 3-methylbutanoate
C7H14O2
MFCD00009203
ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester
CHEBI:31571
CCOC(=O)CC(C)C
Safety and Handling

Safety and Handling

GHS H Statement
H226
Flammable liquid and vapor.

P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c

H226-H315-H319

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.

EINECSNumber : 203-602-3

RTECSNumber : NY1504000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only