Learn More
Ethyl isovalerate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A102920F
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl isovalerate | |
-99°C | |
131°C to 133°C | |
Fruit-like | |
1.396 | |
2500 mL | |
1744677 | |
ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester | |
CCOC(=O)CC(C)C | |
130.187 | |
CHEBI:31571 | |
98% |
108-64-5 | |
0.866 | |
26°C (78°F) | |
C7H14O2 | |
MFCD00009203 | |
UN3272 | |
14,3816 | |
PPXUHEORWJQRHJ-UHFFFAOYSA-N | |
ethyl 3-methylbutanoate | |
7945 | |
130.19 |
Chemical Identifiers
108-64-5 | |
130.187 | |
PPXUHEORWJQRHJ-UHFFFAOYSA-N | |
7945 | |
ethyl 3-methylbutanoate |
C7H14O2 | |
MFCD00009203 | |
ethyl isovalerate, ethyl 3-methylbutyrate, ethylisovalerate, ethyl isopentanoate, isovaleric acid, ethyl ester, butanoic acid, 3-methyl-, ethyl ester, ethyl beta-methylbutyrate, ethyl isovalerianate, 3-methylbutyric acid ethyl ester, 3-methylbutanoic acid ethyl ester | |
CHEBI:31571 | |
CCOC(=O)CC(C)C |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapor.
P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P332+P313-P363-P370+P378q-P501c
H226-H315-H319
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
EINECSNumber : 203-602-3
RTECSNumber : NY1504000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only