missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Chemical Identifiers
CAS | 24066-82-8 |
---|---|
Molecular Formula | C5H7NO2S |
Molecular Weight (g/mol) | 145.176 |
MDL Number | MFCD00060677 |
InChI Key | IYPSSPPKMLXXRN-UHFFFAOYSA-N |
Synonym | Isothiocyanatoacetic Acid Ethyl Ester, Ethoxycarbonylmethyl Isothiocyanate |
PubChem CID | 32202 |
IUPAC Name | ethyl 2-isothiocyanatoacetate |
SMILES | CCOC(=O)CN=C=S |
Chemical Identifiers
24066-82-8 | |
145.176 | |
IYPSSPPKMLXXRN-UHFFFAOYSA-N | |
32202 | |
CCOC(=O)CN=C=S |
C5H7NO2S | |
MFCD00060677 | |
Isothiocyanatoacetic Acid Ethyl Ester, Ethoxycarbonylmethyl Isothiocyanate | |
ethyl 2-isothiocyanatoacetate |
Specifications
215°C | |
5g | |
C5H7NO2S | |
2810 | |
IYPSSPPKMLXXRN-UHFFFAOYSA-N | |
ethyl 2-isothiocyanatoacetate | |
32202 | |
Clear Liquid at 20°C | |
Ethyl Isothiocyanatoacetate |
Undesignated | |
24066-82-8 | |
MFCD00060677 | |
Isothiocyanatoacetic Acid Ethyl Ester, Ethoxycarbonylmethyl Isothiocyanate | |
CCOC(=O)CN=C=S | |
145.176 | |
145.18 | |
≥98.0% (GC) |
Safety and Handling
RTECSNumber : AI5020000
TSCA : Yes