missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Isopropyl Sulfide, TCI America™

Manufacturer:  TCI America E022510G

 View more versions of this product

Catalog No. E022510G



Specifications

Specifications

Ethyl Isopropyl Sulfide
Colorless
5145-99-3
MFCD00039908
NZUQQADVSXWVNW-UHFFFAOYSA-N
2-ethylsulfanylpropane
21228
Clear Liquid at 20°C
103°C
10g
C5H12S
1993
CCSC(C)C
104.211
104.21
≥98.0% (GC)

Chemical Identifiers

5145-99-3
104.211
NZUQQADVSXWVNW-UHFFFAOYSA-N
2-ethylsulfanylpropane
C5H12S
MFCD00039908
21228
CCSC(C)C
Safety and Handling

Safety and Handling

TSCA : No

Documents