missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl isonipecotate, 98+%, ACROS Organics™

$61.11 - $61.11

Chemical Identifiers

CAS 1126-09-6
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.21
MDL Number MFCD00006003
InChI Key RUJPPJYDHHAEEK-UHFFFAOYSA-N
Synonym 4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine, 4-piperidinecarboxylic acid ethyl ester, 4-piperidinecarboxylic acid, ethyl ester, ethyl 4-piperidinecarboxylate, ethyl isonipecotate, ethylpiperidine-4-carboxylate, isonipecotic acid ethyl ester, piperidine-4-carboxylic acid ethyl ester
PubChem CID 70770
IUPAC Name ethyl piperidine-4-carboxylate
SMILES CCOC(=O)C1CCNCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC118640250
SDS View Documents
Thermo Fisher Scientific
118640250
25g Glass bottle
Each for $61.11
 
AC118641000
SDS View Documents
Thermo Fisher Scientific
118641000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

1126-09-6
157.21
RUJPPJYDHHAEEK-UHFFFAOYSA-N
70770
CCOC(=O)C1CCNCC1
C8H15NO2
MFCD00006003
4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine, 4-piperidinecarboxylic acid ethyl ester, 4-piperidinecarboxylic acid, ethyl ester, ethyl 4-piperidinecarboxylate, ethyl isonipecotate, ethylpiperidine-4-carboxylate, isonipecotic acid ethyl ester, piperidine-4-carboxylic acid ethyl ester
ethyl piperidine-4-carboxylate

Specifications

1.0200g/mL
Authentic
1.4585 to 1.4605
95.0°C to 98.0°C (7.0mmHg)
25g
98% min. (GC)
MFCD00006003
4-carbethoxypiperidine, 4-carboethoxypiperidine, 4-ethoxycarbonyl piperidine, 4-piperidinecarboxylic acid ethyl ester, 4-piperidinecarboxylic acid, ethyl ester, ethyl 4-piperidinecarboxylate, ethyl isonipecotate, ethylpiperidine-4-carboxylate, isonipecotic acid ethyl ester, piperidine-4-carboxylic acid ethyl ester
RUJPPJYDHHAEEK-UHFFFAOYSA-N
ethyl piperidine-4-carboxylate
70770
Liquid
Ethyl isonipecotate, 98+%
80°C
Glass bottle
1.02
Brown to Colorless
1126-09-6
C8H15NO2
22, IV, 128
Solubility in water: miscible. Other solubilities: soluble in most organic solvents
CCOC(=O)C1CCNCC1
157.21
157.21
98+%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 214-416-7

SDS
Product Certifications