Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl isonipecotate, 98%

$58.31 - $153.69

Chemical Identifiers

Chemical Name or Material Ethyl isonipecotate
CAS 1126-09-6
Molecular Formula C8H15NO2
Formula Weight 157.21
MDL Number MFCD00006003
Beilstein 118419
Synonym Ethyl piperidine-4-carboxylate; H-DL-Inp-OEt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2060114

  SDS
alfa aesar™
B2060114
25g Each for $58.31

AAB2060122

  SDS
alfa aesar™
B2060122
100g Each for $153.69
Description & Specifications

Specifications

Formula Weight 157.21
Chemical Name or Material Ethyl isonipecotate
Assay 98%
Molecular Formula C8H15NO2
MDL Number MFCD00006003
CAS 1126-09-6
Beilstein 118419
Synonym Ethyl piperidine-4-carboxylate; H-DL-Inp-OEt
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 80°C (176°F)
Boiling Point 203° to 205°C
Quantity 25g

Solubility
Fully miscible with water.

Notes
Store at room temperature. Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.