Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl isodehydroacetate, 98%

$34.11 - $348.80

Chemical Identifiers

Chemical Name or Material Ethyl isodehydroacetate
CAS 3385-34-0
Molecular Formula C10H12O4
Formula Weight 196.2
MDL Number MFCD00006643
Beilstein 145951
Synonym Ethyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate; Isodehydroacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1595506

 
alfa aesar™
A1595506
5g Each for $34.11

AAA1595522

 
alfa aesar™
A1595522
100g Each for $348.80
Description & Specifications

Specifications

Formula Weight 196.2
Chemical Name or Material Ethyl isodehydroacetate
Assay 98%
Molecular Formula C10H12O4
MDL Number MFCD00006643
CAS 3385-34-0
Beilstein 145951
Synonym Ethyl 4,6-dimethyl-2-oxo-2H-pyran-5-carboxylate; Isodehydroacetic acid ethyl ester
Flash Point >110°C (230°F)
Boiling Point 290° to 295°C
Melting Point 17° to 18°C
Quantity 5g

Applications
Ethyl isodehydroacetate is used in the synthesis of bidentate ligands. Also used in the preparation of insect growth regulators with juvenile hormone activity. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong oxidizing agents.