missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl isocyanoacetate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 299550250

 View more versions of this product

Catalog No. AC299550250


 

Specifications Description & Specifications

Specifications

Ethyl isocyanoacetate
25g
1.03g/mL
Glass bottle
Liquid
97%
CNCH2CO2C2H5
Clear amber to dark brown
2999-46-4
Authentic
1.4170 to 1.4250 (20°C, 589nm)
≥96.0% (GC)
C5H7NO2
113.12

Chemical Identifiers

97%
CNCH2CO2C2H5
Ethyl isocyanoacetate
C5H7NO2
113.12
≥96.0% (GC)