Learn More
Ethyl isocyanatoacetate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L1060903
Description
6-(carboxymethylureido)-(±)-nicotine (CMUNic)was prepared by using ethyl isocyanatoacetate. Ethyl isocyanatoacetate has been used in the preparation of nicotine immunogen, ethyl 8-carbamoyl-4-oxo-3,4-dihydroimidazo[5,1-d]-1,2,3,5-tetrazin-3-ylacetate, imidazo[1,2-c]-quinazoline-2,5-(3H,6H)dione.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications6-(carboxymethylureido)-(±)-nicotine (CMUNic)was prepared by using ethyl isocyanatoacetate. Ethyl isocyanatoacetate has been used in the preparation of nicotine immunogen, ethyl 8-carbamoyl-4-oxo-3,4-dihydroimidazo[5,1-d]-1,2,3,5-tetrazin-3-ylacetate, imidazo[1,2-c]-quinazoline-2,5-(3H,6H)dione.
Solubility
Reacts with water.
Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from moisture, water, heat.
Specifications
Ethyl isocyanatoacetate | |
1.151 | |
72°C (161°F) | |
C5H7NO3 | |
OCNCH2CO2CH2CH3 | |
1 g | |
Moisture sensitive | |
Reacts with water. | |
CCOC(=O)CN=C=O | |
129.12 | |
129.12 |
2949-22-6 | |
66°C to 68°C (11 mmHg) | |
Pungent | |
1.424 | |
MFCD00002040 | |
970503 | |
ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester | |
DUVOZUPPHBRJJO-UHFFFAOYSA-N | |
ethyl 2-isocyanatoacetate | |
76283 | |
98% |
Chemical Identifiers
2949-22-6 | |
129.12 | |
DUVOZUPPHBRJJO-UHFFFAOYSA-N | |
76283 | |
CCOC(=O)CN=C=O |
C5H7NO3 | |
MFCD00002040 | |
ethyl isocyanatoacetate, isocyanatoacetic acid ethyl ester, acetic acid, isocyanato-, ethyl ester, carbethoxymethyl isocyanate, isocyanatoethyl acetate, ethoxycarbonylmethyl isocyanate, ethylisocyanatoacetate, ethyl glycolate isocyanate, ethyl n-oxomethylene glycinate, acetic acid,2-isocyanato-, ethyl ester | |
ethyl 2-isocyanatoacetate |
Safety and Handling
GHS H Statement
H334-H302-H335-H227-H315-H319
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P370+P378q-P501c
H227-H302-H315-H317-H319-H334-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 220-967-4
RTECSNumber : AI4450000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only