missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Isobutyrylacetate, TCI America™

Manufacturer:  TCI America E08825G

 View more versions of this product

Catalog No. E08825G



Specifications

Specifications

Ethyl Isobutyrylacetate
Yellow
7152-15-0
MFCD00009198
ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate
CCOC(=O)CC(=O)C(C)C
158.197
158.20
≥95.0% (GC)
90°C
5g
C8H14O3
3272
XCLDSQRVMMXWMS-UHFFFAOYSA-N
ethyl 4-methyl-3-oxopentanoate
81583
Clear Liquid at 20°C

Chemical Identifiers

7152-15-0
158.197
XCLDSQRVMMXWMS-UHFFFAOYSA-N
81583
CCOC(=O)CC(=O)C(C)C
C8H14O3
MFCD00009198
ethyl isobutyrylacetate, 4-methyl-3-oxo-pentanoic acid ethyl ester, pentanoic acid, 4-methyl-3-oxo-, ethyl ester, ethyl isobutyroylacetate, valeric acid, 4-methyl-3-oxo-, ethyl ester, .gamma.,.gamma.-dimethylacetoacetic acid ethyl ester, beta-keto-gamma-methylvaleric acid ethyl ester, ethyl alpha-isobutyroylacetate, ethylisobutyrylacetate, ethyl iso-butyrylacetate
ethyl 4-methyl-3-oxopentanoate
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1486&(2)-1505

TSCA : No

Documents