missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl isobutyrate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 310510025

Catalog No. AC310510025

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Ethyl isobutyrate
97-62-1
112.0°C to 113.0°C
Authentic
Glass bottle
1.3860 to 1.3880
MFCD00009165
0.86
ethyl isobutyrate, ethyl isobutanoate, ethylisobutyrate, isobutyric acid ethyl ester, ethyl 2,2-dimethylacetate, propanoic acid, 2-methyl-, ethyl ester, isobutyric acid, ethyl ester, ethyl 2-methylpropionate, propionic acid, 2-methyl-, ethyl ester, ethyl isobutyrate natural
WDAXFOBOLVPGLV-UHFFFAOYSA-N
ethyl 2-methylpropanoate
7342
116.18
99%
0.8600g/mL
13°C
99%
C6H12O2
(CH3)2CHCO2C2H5
2.5L
15,3869
Solubility in water: not miscible
CCOC(=O)C(C)C
116.18
CHEBI:87303
99%
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly

GHS Signal Word: Danger

EINECSNumber : 202-595-4

RTECSNumber : NQ4675000

TSCA : TSCA

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only