Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl iodoacetate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl iodoacetate
CAS 623-48-3
Molecular Formula ICH2CO2CH2CH3
Formula Weight 214g/mol
MDL Number MFCD00001081
UN Number UN2927
Beilstein 741934
Synonym Iodoacetic acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2417706

 
alfa aesar™
B24177-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2417722

 
alfa aesar™
B24177-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Boiling Point 179° to 180°C
Quantity 5g
Flash Point 76°C (169°F)
Chemical Name or Material Ethyl iodoacetate
CAS 623-48-3
Assay 98%
Molecular Formula ICH2CO2CH2CH3
Formula Weight 214g/mol
MDL Number MFCD00001081
UN Number UN2927
Beilstein 741934
Synonym Iodoacetic acid ethyl ester
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H314-H318-H227
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
RTECS Number AI3575000
TSCA TSCA

Applications
Ethyl iodoacetate was used in the synthesis of trans-2,3-disubstituted indolines by reacting with 1-azido-2-allylbenzene derivatives via a diastereoselective radical cascade.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in dark. Storage temperature is 2-8°C. Store away from oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H314-H318-H227
Fatal if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

2° to 8°C