missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Indoline-2-carboxylate, TCI America™

$447.64 - $1,304.75

Chemical Identifiers

CAS 50501-07-0
Molecular Formula C11H13NO2
Molecular Weight (g/mol) 191.23
MDL Number MFCD06797114
InChI Key KISPUTPAKVZNBI-UHFFFAOYSA-N
Synonym Indoline-2-carboxylic Acid Ethyl Ester
PubChem CID 11949691
IUPAC Name ethyl 2,3-dihydro-1H-indole-2-carboxylate
SMILES CCOC(=O)C1CC2=CC=CC=C2N1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I059525G View Documents TCI America
I059525G
25g Each for $1,304.75
 
I05955G View Documents TCI America
I05955G
5g Each for $447.64
 
Specifications

Chemical Identifiers

50501-07-0
191.23
KISPUTPAKVZNBI-UHFFFAOYSA-N
11949691
CCOC(=O)C1CC2=CC=CC=C2N1
C11H13NO2
MFCD06797114
Indoline-2-carboxylic Acid Ethyl Ester
ethyl 2,3-dihydro-1H-indole-2-carboxylate

Specifications

White-Yellow
25g
C11H13NO2
Indoline-2-carboxylic Acid Ethyl Ester
CCOC(=O)C1CC2=CC=CC=C2N1
191.23
191.23
≥95.0% (GC)
55°C
50501-07-0
MFCD06797114
KISPUTPAKVZNBI-UHFFFAOYSA-N
ethyl 2,3-dihydro-1H-indole-2-carboxylate
11949691
Crystal-Powder at 20°C
Ethyl Indoline-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator