missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl Indoline-2-carboxylate 95.0+%, TCI America™

Supplier:  TCI America I059525G

 View more versions of this product

Catalog No. I059525G



Description

Description

Specifications

Specifications

Ethyl Indoline-2-carboxylate
55°C
C11H13NO2
25 g
KISPUTPAKVZNBI-UHFFFAOYSA-N
ethyl 2,3-dihydro-1H-indole-2-carboxylate
11949691
≥95.0% (GC)
50501-07-0
White-Yellow
MFCD06797114
Indoline-2-carboxylic Acid Ethyl Ester
CCOC(=O)C1CC2=CC=CC=C2N1
191.23
191.23
Crystalline Powder

Chemical Identifiers

50501-07-0
191.23
KISPUTPAKVZNBI-UHFFFAOYSA-N
11949691
CCOC(=O)C1CC2=CC=CC=C2N1
C11H13NO2
MFCD06797114
Indoline-2-carboxylic Acid Ethyl Ester
ethyl 2,3-dihydro-1H-indole-2-carboxylate
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions