missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl indole-3-carboxylate, 97%

$172.72 - $587.55

Chemical Identifiers

CAS 776-41-0
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.214
MDL Number MFCD00228454
InChI Key XOUHVMVYFOXTMN-UHFFFAOYSA-N
Synonym ethyl indole-3-carboxylate, indole-3-carboxylic acid ethyl ester, 1h-indole-3-carboxylic acid ethyl ester, 1h-indole-3-carboxylic acid, ethyl ester, indole3carboxylicacidethylester, pubchem7352, lindole-3-ethyl formate, 3-ethoxycarbonyl indole, ksc644o3d, 3-indolecarboxylicacidethylester
PubChem CID 247965
IUPAC Name ethyl 1H-indole-3-carboxylate
SMILES CCOC(=O)C1=CNC2=CC=CC=C21
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6625403 View Documents Alfa Aesar™
H6625403
1g Each for $172.72
 
AAH6625406 View Documents Alfa Aesar™
H6625406
5g Each for $587.55
 
Specifications

Chemical Identifiers

776-41-0
189.214
XOUHVMVYFOXTMN-UHFFFAOYSA-N
247965
CCOC(=O)C1=CNC2=CC=CC=C21
C11H11NO2
MFCD00228454
ethyl indole-3-carboxylate, indole-3-carboxylic acid ethyl ester, 1h-indole-3-carboxylic acid ethyl ester, 1h-indole-3-carboxylic acid, ethyl ester, indole3carboxylicacidethylester, pubchem7352, lindole-3-ethyl formate, 3-ethoxycarbonyl indole, ksc644o3d, 3-indolecarboxylicacidethylester
ethyl 1H-indole-3-carboxylate

Specifications

120°C to 124°C
776-41-0
MFCD00228454
ethyl indole-3-carboxylate, indole-3-carboxylic acid ethyl ester, 1h-indole-3-carboxylic acid ethyl ester, 1h-indole-3-carboxylic acid, ethyl ester, indole3carboxylicacidethylester, pubchem7352, lindole-3-ethyl formate, 3-ethoxycarbonyl indole, ksc644o3d, 3-indolecarboxylicacidethylester
CCOC(=O)C1=CNC2=CC=CC=C21
189.214
189.21
97%
1g
C11H11NO2
147476
XOUHVMVYFOXTMN-UHFFFAOYSA-N
ethyl 1H-indole-3-carboxylate
247965
Solid
Ethyl indole-3-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning