missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl indole-3-acrylate, 97%, Thermo Scientific Chemicals
$83.84 - $267.64
Chemical Identifiers
CAS | 15181-86-9 |
---|---|
Molecular Formula | C13H13NO2 |
Molecular Weight (g/mol) | 215.252 |
MDL Number | MFCD00051947 |
InChI Key | OQJSITNIWIYWPU-FPLPWBNLSA-N |
Synonym | ethyl 2z-3-1h-indol-3-yl prop-2-enoate |
PubChem CID | 13863197 |
IUPAC Name | ethyl (Z)-3-(1H-indol-3-yl)prop-2-enoate |
SMILES | CCOC(=O)C=CC1=CNC2=CC=CC=C21 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0870303
|
Thermo Scientific Chemicals
L0870303 |
1 g |
Each for $83.84
|
|
|||||
AAL0870306
|
Thermo Scientific Chemicals
L0870306 |
5 g |
Each for $267.64
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
15181-86-9 | |
215.252 | |
OQJSITNIWIYWPU-FPLPWBNLSA-N | |
13863197 | |
CCOC(=O)C=CC1=CNC2=CC=CC=C21 |
C13H13NO2 | |
MFCD00051947 | |
ethyl 2z-3-1h-indol-3-yl prop-2-enoate | |
ethyl (Z)-3-(1H-indol-3-yl)prop-2-enoate |
Specifications
15181-86-9 | |
C13H13NO2 | |
1 g | |
ethyl 2z-3-1h-indol-3-yl prop-2-enoate | |
CCOC(=O)C=CC1=CNC2=CC=CC=C21 | |
215.252 | |
215.25 | |
Ethyl indole-3-acrylate |
118°C to 122°C | |
MFCD00051947 | |
476317 | |
OQJSITNIWIYWPU-FPLPWBNLSA-N | |
ethyl (Z)-3-(1H-indol-3-yl)prop-2-enoate | |
13863197 | |
97% |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only