missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl indole-2-carboxylate, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 118600050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Ethyl indole-2-carboxylate | |
3770-50-1 | |
96% min. (GC) | |
C11H11NO2 | |
5g | |
ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole | |
QQXQAEWRSVZPJM-UHFFFAOYSA-N | |
ethyl 1H-indole-2-carboxylate | |
73125 | |
97% |
97% | |
Authentic | |
Glass bottle | |
MFCD00005609 | |
22, I, 506 | |
Solubility in water: insoluble in water | |
CCOC(=O)C1=CC2=CC=CC=C2N1 | |
189.21 | |
189.21 |
Chemical Identifiers
3770-50-1 | |
189.21 | |
QQXQAEWRSVZPJM-UHFFFAOYSA-N | |
73125 | |
CCOC(=O)C1=CC2=CC=CC=C2N1 |
C11H11NO2 | |
MFCD00005609 | |
ethyl indole-2-carboxylate, indole-2-carboxylic acid ethyl ester, 2-carbethoxyindole, 1h-indole-2-carboxylic acid, ethyl ester, indole-2-carboxylic acid, ethyl ester, 1h-indole-2-carboxylic acid ethyl ester, ethylindole-2-carboxylate, ethyl 2-indolecarboxylate, 1h-indole-2-carboxylic acid,ethyl ester, 2-carboethoxyindole | |
ethyl 1H-indole-2-carboxylate |
Safety and Handling
EINECSNumber : 223-206-4
RUO â Research Use Only