missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl imidazole-4-carboxylate, 98%

Manufacturer:  Alfa Aesar™ H6409503

 View more versions of this product

Catalog No. AAH6409503



Description

Description

Applications
A ring-substituted-1H-imidazole-4-carboxylic acid derivative, that is shown to act as a new class of anti-tuberculosis agents.

Solubility
Soluble in dimethyl sulfoxide and methanol. Slightly soluble in water.

Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Specifications

Ethyl imidazole-4-carboxylate
1g
C6H8N2O2
606937
KLWYPRNPRNPORS-UHFFFAOYSA-N
ethyl 1H-imidazole-5-carboxylate
99170
Solid
156°C to 160°C
23785-21-9
MFCD02646639
ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester
CCOC(=O)C1=CN=CN1
140.142
140.14
98%

Chemical Identifiers

23785-21-9
140.142
KLWYPRNPRNPORS-UHFFFAOYSA-N
99170
CCOC(=O)C1=CN=CN1
C6H8N2O2
MFCD02646639
ethyl 1h-imidazole-4-carboxylate, ethyl imidazole-4-carboxylate, ethyl 4-imidazolecarboxylate, ethyl 4 5-imidazolecarboxylate, 4-carbethoxyimidazole, 1h-imidazole-4-carboxylic acid ethyl ester, nic7lr0zon, unii-nic7lr0zon, imidazole-4-carboxylic acid ethyl ester, 1h-imidazole-4-carboxylic acid, ethyl ester
ethyl 1H-imidazole-5-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

SDS Documents