missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Ethyl hydrogen suberate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0427206
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Ethyl hydrogen suberate | |
25°C to 27°C | |
MFCD00014385 | |
1775737 | |
KCFVQEAPUOVXTH-UHFFFAOYSA-N | |
8-ethoxy-8-oxooctanoic acid | |
84204 | |
98% |
14113-01-0 | |
C10H18O4 | |
5 g | |
ethyl hydrogen suberate, octanedioic acid, ethyl, korksaureathylester, acmc-1b2e5, 8-ethoxy-8-oxo-octanoic acid, octanedioic acid 1-ethyl ester, octanedioic acid,1-ethyl ester, octanedioic acid, 1-ethyl ester, 8-ethoxy-8-oxidanylidene-octanoic acid, ethylhydrogensuberate, monoethylsuberate; subericacid monoethylester | |
CCOC(=O)CCCCCCC(=O)O | |
202.25 | |
202.25 |
Chemical Identifiers
14113-01-0 | |
202.25 | |
KCFVQEAPUOVXTH-UHFFFAOYSA-N | |
8-ethoxy-8-oxooctanoic acid |
C10H18O4 | |
MFCD00014385 | |
84204 | |
CCOC(=O)CCCCCCC(=O)O |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 237-968-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only