Learn More
Ethyl hydrogen pimelate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0402514
Description
Ethyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsEthyl hydrogen pimelate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Solubility
Not miscible or difficult to mix with water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Ethyl hydrogen pimelate | |
10°C | |
135°C to 138°C (1 mmHg) | |
C9H16O4 | |
CH3CH2O2C(CH2)5CO2H | |
25 g | |
ethyl hydrogen pimelate, pimelic acid monoethyl ester, heptanedioic acid monoethyl ester, ethyl hydrogen heptane-1,7-dioate, ethyl hydroden pimelate, heptandioic acid monoethyl ester, heptanedioic acid 1-ethyl ester, heptanedioic acid, 1-ethyl ester | |
NQYXFXWKKYGBNL-UHFFFAOYSA-N | |
7-ethoxy-7-oxoheptanoic acid | |
118383 | |
98% |
33018-91-6 | |
1.06 | |
>110°C (230°F) | |
1.442 | |
MFCD00801202 | |
1774583 | |
Not miscible or difficult to mix with water. | |
CCOC(=O)CCCCCC(=O)O | |
188.223 | |
188.22 |
Chemical Identifiers
33018-91-6 | |
188.223 | |
NQYXFXWKKYGBNL-UHFFFAOYSA-N | |
118383 | |
CCOC(=O)CCCCCC(=O)O |
C9H16O4 | |
MFCD00801202 | |
ethyl hydrogen pimelate, pimelic acid monoethyl ester, heptanedioic acid monoethyl ester, ethyl hydrogen heptane-1,7-dioate, ethyl hydroden pimelate, heptandioic acid monoethyl ester, heptanedioic acid 1-ethyl ester, heptanedioic acid, 1-ethyl ester | |
7-ethoxy-7-oxoheptanoic acid |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
EINECSNumber : 251-346-6
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only