missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ethyl hydrogen malonate, 90+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 04469236

 View more versions of this product

Catalog No. AA4469236


Description

Description

Ethyl hydrogen malonate is used as a reactant for the preparation of tetramic acids through Dieckmann ring closure and organocatalytic decarboxylative Doebner-Knoevenagel reactions. It is involved in the acylation reactions and Knoevenagel condensation with aldehydes. It is also used in the preparation of gamma-lactones from olefins by intermolecular carbolactonization in presence of Mn(III) acetate as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Ethyl hydrogen malonate
Yellow
106°C (3 mmHg)
C5H8O4
HO2CCH2CO2CH2CH3
500 g
ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate
HGINADPHJQTSKN-UHFFFAOYSA-N
3-ethoxy-3-oxopropanoic acid
70615
132.12
Liquid
1071-46-1
1.119 g/mL
>110°C (230°F)
1.4295
MFCD00020490
1758845
Miscible with water,chloroform and other solvents.
CCOC(=O)CC(=O)O
132.115
CHEBI:86907
≥90%

Chemical Identifiers

1071-46-1
132.115
HGINADPHJQTSKN-UHFFFAOYSA-N
70615
3-ethoxy-3-oxopropanoic acid
C5H8O4
MFCD00020490
ethyl hydrogen malonate, monoethyl malonate, monoethyl malonic acid, mono-ethyl malonate, monoethyl hydrogen malonate, malonic acid monoethyl ester, ethoxycarbonyl acetic acid, 2-ethoxycarbonyl acetic acid, 3-ethoxy-3-oxo-propanoic acid, ethylhydrogenmalonate
CHEBI:86907
CCOC(=O)CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 213-992-7

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only