Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl hydrogen glutarate, 97%

$458.05 - $458.05

Chemical Identifiers

Chemical Name or Material Ethyl hydrogen glutarate
CAS 1070-62-8
Molecular Formula C7H12O4
Formula Weight 160.17
MDL Number MFCD00014379
Beilstein 1766397
Synonym Glutaric acid monoethyl ester; Monoethyl glutarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0394114

  SDS
alfa aesar™
L0394114
25g Each for $458.05
Description & Specifications

Specifications

Formula Weight 160.17
Molecular Formula C7H12O4
MDL Number MFCD00014379
Assay 97%
CAS 1070-62-8
Beilstein 1766397
Synonym Glutaric acid monoethyl ester; Monoethyl glutarate
Boiling Point 118° to 120°C (1.5mmHg)
Health Hazard 1 Warning
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Ethyl hydrogen glutarate
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.