Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl hydrogen fumarate, 97%

$61.78 - $726.26

Chemical Identifiers

Chemical Name or Material Ethyl hydrogen fumarate
CAS 2459-05-4
Molecular Formula C6H8O4
Formula Weight 144.13
MDL Number MFCD00002699
Synonym Fumaric acid monoethyl ester; Monoethyl fumarate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1254514

  SDS
alfa aesar™
A1254514
25g Each for $61.78

AAA1254522

  SDS
alfa aesar™
A1254522
100g Each for $167.26

AAA1254536

  SDS
alfa aesar™
A1254536
500g Each for $726.26
Description & Specifications

Specifications

Formula Weight 144.13
Chemical Name or Material Ethyl hydrogen fumarate
Assay 97%
Molecular Formula C6H8O4
MDL Number MFCD00002699
CAS 2459-05-4
Health Hazard 1 Warning
Melting Point 64° to 65°C
Quantity 25g
Synonym Fumaric acid monoethyl ester; Monoethyl fumarate
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
Ethyl hydrogen fumarate was used in the preparation of photo-crosslinkable macromers. It was also used to synthesize Ugi/intramolecular Diels-Alder (IMDA) cycloaddition products.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.