Please login to your online account to display your discounted pricing

Alfa Aesar™ Ethyl heptafluorobutyrate, 98%

Chemical Identifiers

Chemical Name or Material Ethyl heptafluorobutyrate
CAS 356-27-4
Molecular Formula CF3CF2CF2CO2CH2CH3
Formula Weight 242.09
MDL Number MFCD00000434
UN Number UN3272
Beilstein 1794634
Synonym Heptafluorobutyric acid ethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1674214

 
alfa aesar™
A1674214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1674222

 
alfa aesar™
A1674222
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.09
Chemical Name or Material Ethyl heptafluorobutyrate
TSCA TSCA
Assay 98%
Molecular Formula CF3CF2CF2CO2CH2CH3
MDL Number MFCD00000434
CAS 356-27-4
UN Number UN3272
Beilstein 1794634
Synonym Heptafluorobutyric acid ethyl ester
Boiling Point 95° to 98°C
Health Hazard 1 Danger
Quantity 25g
Flash Point 14°C (57°F)
Health Hazard 2 GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ESTERS, N.O.S.
RTECS Number ET4200000
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.