Please login to your online account to display your discounted pricing

Ethyl heptafluorobutyrate, 97%, ACROS Organics™

$325.92 - $325.92

Chemical Identifiers

Chemical Name or Material Ethyl heptafluorobutyrate
Name Note 97%
CAS 356-27-4
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H5F7O2
Linear Formula CF3CF2CF2CO2C2H5
Formula Weight 242.09
MDL Number MFCD00000434
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC174441000

  SDS
thermo fisher scientific
174441000
Glass bottle 100g Each for $325.92
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 242.09
Quantity 100g
Color Clear colorless
EINECS Number 206-602-1
TSCA TSCA
RTECS Number ET4200000
Chemical Name or Material Ethyl heptafluorobutyrate
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C6H5F7O2
Refractive Index 1.3010 to 1.3030 (20°C, 589nm)
Linear Formula CF3CF2CF2CO2C2H5
MDL Number MFCD00000434
CAS 356-27-4
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.