Please login to your online account to display your discounted pricing

Ethyl glycidyl ether, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Ethyl glycidyl ether
Name Note 98%
CAS 773168
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C5H10O2
Formula Weight 102.13
MDL Number MFCD00085264
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC299230250

  SDS
thermo fisher scientific
299230250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC299230050

  SDS
thermo fisher scientific
299230050
Glass bottle 5mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 102.13
Quantity 25mL
Color Clear colorless
Density 0.94g/mL
EINECS Number 223-671-3
TSCA TSCA
RTECS Number TZ3200000
CAS 773168
Chemical Name or Material Ethyl glycidyl ether
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C5H10O2
Refractive Index 1.4075 to 1.4095 (20°C, 589nm)
MDL Number MFCD00085264
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.